Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione S Transferase kappa 1 Antibody (SR1382), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322046100UL
Description
Glutathione S Transferase kappa 1 Monoclonal antibody specifically detects Glutathione S Transferase kappa 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
Glutathione S Transferase kappa 1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR1382 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
EC 2.5.1.18, glutathione S-transferase k1, glutathione S-transferase kappa 1, Glutathione S-transferase subunit 13, glutathione S-transferase subunit 13 homolog, GST, GST 13-13, GST class-kappa, GST13, GST13-13, GSTK1-1, hGSTK1 | |
A synthesized peptide derived from human Glutathione S Transferase kappa 1 (Uniprot #: Q9Y2Q3) | |
100 μg | |
Endocrinology | |
373156 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction