Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione S Transferase kappa 1 Rabbit anti-Human, Mouse, Clone: 7J5B3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Glutathione S Transferase kappa 1 |
---|---|
Clone | 7J5B3 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Glutathione S Transferase kappa 1 Monoclonal antibody specifically detects Glutathione S Transferase kappa 1 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Glutathione S Transferase kappa 1 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Endocrinology | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
373156 | |
IgG | |
Affinity purified |
7J5B3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
EC 2.5.1.18, glutathione S-transferase k1, glutathione S-transferase kappa 1, Glutathione S-transferase subunit 13, glutathione S-transferase subunit 13 homolog, GST, GST 13-13, GST class-kappa, GST13, GST13-13, GSTK1-1, hGSTK1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human Glutathione S Transferase kappa 1 (Q9Y2Q3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title