Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione S-Transferase mu 1/GSTM1 Antibody (1H7.D8.G5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP244180
Description
Ensure accurate, reproducible results in Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Glutathione S-Transferase mu 1/GSTM1 Monoclonal specifically detects Glutathione S-Transferase mu 1/GSTM1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation.Specifications
Glutathione S-Transferase mu 1/GSTM1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:1000, ELISA 1:50000-1:200000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500 | |
P09488, P09488, P09488, P09488 | |
GSTM1 | |
This Protein A purified Glutathione S-Transferase mu 1/GSTM1 Antibody (1H7.D8.G5) was prepared by repeated immunizations in mice with Glutathione S-Transferase mu 1/GST Glutathione S-Transferase mu 1/GSTM1 Protein (Uniprot: P09488) | |
Protein A purified | |
RUO | |
Primary | |
Reacts specifically with Glutathione S-Transferase mu 1/GSTM1 protein. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
1H7.D8.G5 | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase | |
Mouse | |
25.71 kDa | |
0.1 mg | |
Asthma, Cancer, Immunology | |
2944 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction