Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione S-transferase Mu 5 Antibody, Alexa Fluor™ 405 , Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297064AF405
Description
Glutathione S-transferase Mu 5 Polyclonal antibody specifically detects Glutathione S-transferase Mu 5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Glutathione S-transferase Mu 5 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
EC 2.5.1.18, glutathione S-alkyltransferase M5, glutathione S-aralkyltransferase M5, glutathione S-aryltransferase M5, glutathione S-transferase M5, glutathione S-transferase mu 5, GST class-mu 5, GSTM5-5, GTM5, S-(hydroxyalkyl)glutathione lyase M5 | |
Produced in rabbits immunized with purified, recombinant Human Glutathione S-transferase Mu 5 (AAH58881.1; Met1-Lys218). | |
0.1 mL | |
Signal Transduction | |
2949 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction