Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione S-Transferase pi 1/GSTP1 Antibody (028), DyLight 594

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289324DL594
Description
Glutathione S-Transferase pi 1/GSTP1 Monoclonal specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human samples. It is validated for Western Blot.Specifications
Glutathione S-Transferase pi 1/GSTP1 | |
Monoclonal | |
DyLight 594 | |
deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI | |
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the Human Glutathione S-Transferase pi 1/GSTP1. | |
0.1 mL | |
Asthma, Cancer, Immunology, Neuroscience | |
2950 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
28 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction