Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Glutathione Sepharose™ 4B Media
GSA_VA

Catalog No. 45000140 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
300 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-140 300 mL
45-000-139 10 mL
45-002-065 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-000-140 Supplier Cytiva Supplier No. 17075604
Add to Cart
Add to Cart
Request a Quote

Use for high capacity purification of Glutathione S-Transferase (GST) tagged fusion proteins. Cytiva Life Sciences™ Glutathione Sepharose™ 4B Media requires only a single-step for purification.

  • Binding capacity: >5mg horse liver GST/mL medium (flowrate dependent) Average particle size: 90μm
  • Recommended flowrate (H2O at room temp.): 75cm/hour
  • Stable in presence of all commonly used aqueous buffers
  • Stable in pH range 4 to 13

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Material Glutathione Sepharose 4 B
pH Range 4 to 13
Particle Size 90 μm
Quantity 300 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.