Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione Synthetase Antibody (12F2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259455H
This item is not returnable.
View return policy
Description
Glutathione Synthetase Monoclonal antibody specifically detects Glutathione Synthetase in Human samples. It is validated for Western Blot, ELISASpecifications
Glutathione Synthetase | |
Monoclonal | |
HRP | |
EC 6.3.2.3, Glutathione synthase, glutathione synthetase, GSH synthetase, GSHS, GSH-S, MGC14098 | |
Recombinant human Glutathione Synthetase (1-474aa) purified from E. coli | |
0.1 mL | |
Neuroscience | |
2937 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
12F2 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction