Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione Synthetase Rabbit anti-Human, Mouse, Rat, Clone: 2S9W7, Novus Biologicals™

Rabbit Monoclonal Antibody
$208.87 - $463.77
Specifications
| Antigen | Glutathione Synthetase |
|---|---|
| Clone | 2S9W7 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Glutathione Synthetase Monoclonal antibody specifically detects Glutathione Synthetase in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Glutathione Synthetase | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2937 | |
| IgG | |
| Affinity purified |
| 2S9W7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 6.3.2.3, Glutathione synthase, glutathione synthetase, GSH synthetase, GSHS, GSH-S, MGC14098 | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human Glutathione Synthetase (GSS) (P48637). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title