Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Gly-Phe-7-Amino-4-Trifluoromethylcoumarin
Gly-Phe-7-Amino-4-Trifluoromethylcoumarin, White powder
$335.00 - $638.00
Specifications
Color | White |
---|---|
Form | Powder |
For Use With (Application) | Cell Migration Analysis; Cell Signaling Analysis |
Molecular Weight (g/mol) | 433.38 ; 574 (Including TFA salt) |
Product Type | Proteins, Enzymes & Peptides |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP3AFC03310
|
MP Biomedicals™
03AFC03310 |
10mg |
Each for $335.00
|
|
|||||
MP3AFC03325
|
MP Biomedicals™
03AFC03325 |
25mg |
Each for $638.00
|
|
|||||
Description
- Gly-Phe-AFC is a fluorogenic, cell-permeant peptide substrate, used in Cytotoxicity testing.
- In vitro cytotoxicity testing has become an integral aspect of drug discovery because it is a convenient, costeffective, and predictive means of characterizing the toxic potential of new chemical entities.
- It is also used as substrate for measuring Dipeptidyl peptidase1 (DPP1) activity.
Specifications
White | |
Cell Migration Analysis; Cell Signaling Analysis | |
Proteins, Enzymes & Peptides |
Powder | |
433.38 ; 574 (Including TFA salt) | |
Keep cool and dry |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title