Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLYAT Antibody (OTI4D1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271564H
Description
GLYAT Monoclonal specifically detects GLYAT in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
GLYAT | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
GLYAT | |
Full length recombinant protein of human GLYAT (NP_005829) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI4D1 | |
Western Blot, Immunohistochemistry | |
ACGNATAAc, Acyl-CoA:glycine N-acyltransferase, Aralkyl acyl-CoA N-acyltransferase, Aralkyl acyl-CoA:amino acid N-acyltransferase, aralkyl-CoA N-acyltransferase, CATEC 2.3.1.13, GATHRP-1(CLP), glycine N-acyltransferase, glycine-N-acyltransferase | |
Mouse | |
Affinity Purified | |
metabolism | |
10249.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction