Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLYAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589468 Supplier Invitrogen™ Supplier No. PA589468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMLPLQGAQM LQMLEKSLRK SLPASLKVYG TVFHINHGNP FNLKAVVDKW PDFNTVVVCP QEQDMTDDLD HYTNTYQIYS KDPQNCQEFL GSPELINWKQ HLQIQSSQPS LNEAIQNLAA IKSFKVKQTQ RILYMAAETA KELTPFLLKS KILSPNGGKP KAM; Positive Samples: 293T; Cellular Location: Mitochondrion.

Mitochondrial acyltransferase which transfers an acyl group to the N-terminus of glycine and glutamine, although much less efficiently. Can conjugate numerous substrates to form a variety of N-acylglycines, with a preference for benzoyl-CoA over phenylacetyl-CoA as acyl donors. Thereby detoxify xenobiotics, such as benzoic acid or salicylic acid, and endogenous organic acids, such as isovaleric acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLYAT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GLYAT
Gene Accession No. Q6IB77
Gene Alias A330009E03Rik; AAc; Ab2-132; ACGNAT; Acyl-CoA:glycine N-acyltransferase; AI195249; AI315345; Aralkyl acyl-CoA N-acyltransferase; aralkyl acyl-CoA:amino acid N-acyltransferase; aralkyl-CoA N-acyltransferase; benzoyl-coenzyme A:glycine N-acyltransferase; CAT; GAT; Glyat; Glycine N-acyltransferase; glycine N-benzoyltransferase; glycine-N-acyltransferase; HRP-1(CLP); Liver regeneration-related protein LRRG067
Gene Symbols GLYAT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-163 of human GLYAT (NP_0058293).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10249
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.