Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLYATL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543275
Description
Peptide sequence: LDEAIRKVAT SKSVQVDYMK TILFIPELPK KHKTSSNDKM ELFEVDDDNK Sequence homology: Human: 100%.
GLYATL2 belongs to the glycine N-acyltransferase family. GLYATL2 is a mitochondrial acyltransferase which transfers the acyl group to the N-terminus of glycine. It can conjugate a multitude of substrates to form a variety of N-acylglycines.
Specifications
GLYATL2 | |
Polyclonal | |
Unconjugated | |
GLYATL2 | |
acyl-CoA:glycine N-acyltransferase-like protein 2; BXMAS2-10; GATF-B; GLYATL2; glycine acyltransferase family-B; glycine N-acyltransferase-like protein 2; glycine-N-acyltransferase like 2; glycine-N-acyltransferase-like 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
219970 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WU03 | |
GLYATL2 | |
Synthetic peptide directed towards the middle region of human GLYATL2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction