Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLYATL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569588
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Rat: 93%.
Catalyzes the conjugation of long-chain fatty acyl-CoA thioester and glycine to produce long-chain N-(fatty acyl)glycine, an intermediate in the primary fatty acid amide biosynthetic pathway. [UniProt].
Specifications
GLYATL3 | |
Polyclonal | |
Unconjugated | |
GLYATL3 | |
Acyl-CoA:glycine-N-acyltransferase-like protein 3; bA28H17.2; C6orf140; EG435528; GLYATL3; Glycine N-acyltransferase-like protein 3; glycine N-acyltransferase-like protein 3-like; glycine-N-acyltransferase like 3; glycine-N-acyltransferase-like 3; Glycine-N-acyltransferase-like protein 3; Gm5683; LOW QUALITY PROTEIN: glycine N-acyltransferase-like protein 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
389396 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5SZD4 | |
GLYATL3 | |
synthetic peptide directed towards the N terminal region of human GLYATL3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction