Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Glybenclamide (NLRP3 Inhibitor)
SDP

Catalog No. NBP230141 Shop All Novus Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP230141 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP230141 Supplier R&D Systems™ Supplier No. NBP230141

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Glybenclamide, also known as glyburide, is an ATP-sensitive potassion channel inhibitor that has been shown to inhibit various biological processes including PAMP and DAMP-induced activation of the NRLP3 inflammasome as well as the activation of the inflammasome by crystalline substances

Form : White Solid Reconstitute: Dilute further with sterilized PBS. Make a stock solution of 25mg/mL of the NLRP3 inhibitor in high purity DMSO (>99.9%).

Specifications

For Use With (Application) Inhibition of NLRP3
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 1000 mg
Product Type Glybenclamide
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.