Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyburide, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602945
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle

G1130, 10238-21-8, C23H28ClN3O5S

Spectrum™ Chemical Glyburide, USP is an oral diabetes medicine that helps control blood sugar levels and is used to treat type 2 diabetes. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10238-21-8
Molecular Formula C23H28ClN3O5S
Molecular Weight (g/mol) 494.00
InChI Key ZNNLBTZKUZBEKO-UHFFFAOYSA-N
IUPAC Name 5-chloro-N-[2-(4-{[(cyclohexylcarbamoyl)amino]sulfonyl}phenyl)ethyl]-2-methoxybenzamide
SMILES COC1=CC=C(Cl)C=C1C(=O)NCCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.