Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GlycanPac™ AXR-1 HPLC Columns
GREENER_CHOICE

Catalog No. dx088136
Change view
Click to view available options
Length (Metric):
150 mm
250 mm
Diameter (Metric):
2.1 mm
Particle Size:
1.9 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size
DX088136 150 mm 2.1 mm 1.9 μm
DX088135 250 mm 2.1 mm 1.9 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DX088136 Supplier Thermo Scientific™ Supplier No. 088136
Add to Cart
Add to Cart

Get ultra-high resolution and selectivity for both labeled and native glycans using this HPLC/UHPLC column. It's compatible with MS and fluorescence detection.

Get industry-leading, ultra-high resolution of labeled and native glycans for structural characterization and quantification using the Thermo Scientific™ GlycanPac™ AXR-1 LC Columns. This high-performance, silica-based chromatography column achieves mixed-mode separation using a combination of charge, isomer structure, and size. Now there's a column with unique selectivity to analyze biologically important glycans. The GlycanPac AXR-1 column is available in multiple HPLC and UHPLC formats and is compatible with fluorescence or mass spectrometry (MS) detection.

Exceptional Glycan Selectivity with Industry-Leading Resolution

Ultra-high resolution and selectivity of glycans are attributed to the GlycanPac AXR-1 column's innovative mixed-mode surface chemistry, which combines:

  • WAX functionality for retention of negatively charged glycans, with separation in order of increasing charge
  • Reversed-phase functionality for same-charge glycans, with separation according to isomerism and size
Reliable Column Performance

This high-purity spherical silica column is designed and tested for HPLC and UHPLC methods using either fluorescence or MS detection applications. The GlycanPac AXR-1 column uses volatile aqueous buffers (e.g., ammonium acetate or ammonium formate) and acetonitrile, presenting the eluting glycans ready for introduction into MS instruments.

Available in Multiple Formats
  • Use 1.9μm columns for UHPLC applications and 3μm columns for HPLC applications
  • Select from a variety of dimensions to support the resolution, throughput, or instrumental requirements required for your application
  • Protect the analytical column with GycanPac AXR-1 Guard columns
TRUSTED_SUSTAINABILITY

Specifications

Column Chemistry WAX and RP Mixed Mode
Column Format Analytical Column
Diameter (Metric) 2.1 mm
Flow Rate 0.2 to 0.4 mL/min
Length (Metric) 150 mm
Max. Pressure 10000 psi (690 bar)
Particle Shape Spherical, high purity, porous
Particle Size 1.9 μm
pH 2 to 8
Pore Size 175 Å
Quantity Each
Solvents Compatible with 0 to 100% aqueous and common HPLC solvents
Surface Area 220 m²/g
Temperature 60°C
Stationary Phase Proprietary
Product Line GlycanPac
Packing Material Spherical, Fully Porous, Ultrapure Silica
Column Type Mixed Mode
Phase Reversed Phase and Ion Exchange
Technique Bio-LC
Green Features FSC
Product Type Analytical Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.