Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RSOG002020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOG002020F 20 L Cubitainer
RSOG0020500 500 mL Natural Poly Bottle
RSOG00201A 1 L Natural Poly Bottle
RSOG00204A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RSOG002020F Supplier Ricca Chemical Company Supplier No. RSOG002020F
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

CAS Max % 100
Boiling Point 544°C
Ignition Residue max. 0.005%
Packaging Cubitainer
Quantity 20 L
Percent Purity min. 99.5%
Grade ACS
Physical Form Liquid
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.