Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin, MP Biomedicals™

Catalog No. MP219956604 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219956604 4 L
MP21995665 500 mL
MP219956601 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219956604 Supplier MP Biomedicals, Inc Supplier No. 0219956604
Add to Cart
Add to Cart

Used in sample prep and gel formation for polyacrylamide gel electrophoresis. Glycerin, MP Biomedicals (5-10%) increases the density of a sample so that it will layer at the bottom of a sample well. It is also used in casting gradient gels and as a protei

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 20°C (literature)
Density 1.25g/mL (Lit.)
Boiling Point 182°C (20 mmHg (literature))
Ignition Residue ≤0.01%
Refractive Index n20/D 1.474 (Lit.)
Quantity 4 L
Specific Gravity ≥1.249
Vapor Density 3.1 (vs air) (Lit.)
Vapor Pressure <1mmHg (20°C) (Lit.)
Formula Weight 92.09g/mol
Percent Purity ≥99%
Grade USP
Physical Form Liquid
Show More Show Less
Recommended Storage Store at Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.