Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycerol-1-allylether, 98%, Thermo Scientific™
Click to view available options
:
100mL; Glass bottle
Specifications
Specifications
CAS | 123-34-2 |
Density | 1.0700g/mL |
Boiling Point | 245.0°C |
Flash Point | 132°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C6H12O3 |
Linear Formula | CH2=CHCH2OCH(OH)CH2OH |
Refractive Index | 1.4610 to 1.4630 |
Show More |
Safety and Handling
Hazard Statement | GHS P Statement Causes serious eye irritation. Causes skin irritation. |
Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction