Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycerol 3 Phosphate Dehydrogenase Antibody (104), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290308FR
Description
Glycerol 3 Phosphate Dehydrogenase Monoclonal specifically detects Glycerol 3 Phosphate Dehydrogenase in Human samples. It is validated for ELISA.Specifications
Glycerol 3 Phosphate Dehydrogenase | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
2819 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
104 | |
ELISA | |
EC 1.1.1, EC 1.1.1.8, FLJ26652, glycerol-3-phosphate dehydrogenase [NAD+], cytoplasmic, glycerol-3-phosphate dehydrogenase 1 (soluble), glycerol-3-phosphate dehydrogenase, soluble, GPD-C, GPDH-C | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Glycerol 3 Phosphate Dehydrogenase (P21695; Met1-Met349). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction