Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014656 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL

Catalog No. 11-101-4656

Supplier: MilliporeSigma Supelco 770671ML

Only null left
Add to Cart
Add to Cart

Glycerol is a simple polyol compound, which is obtained as a byproduct of soap and biodiesel industries.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 698°F
CAS 56-81-5
Vapor Pressure <1 mmHg (20°C)
Vapor Density 3.1 (vs. air)
Density 1.25 g/mL
Flash Point 199°C - Pensky-Martens°Closed°Cup
Physical Form Neat
Linear Formula HOCH2 CH(OH)CH2 OH
MDL Number MFCD00004722
Melting Point 20°C
Boiling Point 182°C (20 mmHg)
Synonym 1,2,3-Propanetriol; Glycerin
Type Analytical Standard
Quantity 1 mL
Formula Weight 92.09
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.