Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575254
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
GK2 (glycerol kinase 2), also known as GKP2 or GKTA, is a 553 amino acid protein that belongs to the FGGY kinase family and is involved in the pathway of glycerol degradation. Localized to the outer membrane of the mitochondrion and expressed at high levels in testis, GK2 functions to catalyze the ATPdependent conversion of glycerol to glycerol 3-phosphate. Via its catalytic activity, GK2 plays an essential role in the regulation of glycerol uptake and metabolism.
Specifications
GK2 | |
Polyclonal | |
Unconjugated | |
GK2 | |
ATP:glycerol 3-phosphotransferase 2; GK; GK 2; GK2; GKP2; Gkrs2; Gk-rs2; GKTA; glucokinase activity, related sequence 2; Glycerokinase; glycerokinase 2; glycerol kinase 2; glycerol kinase pseudogene 2; glycerol kinase testis specific 2; glycerol kinase, testis specific 2; testis tissue sperm-binding protein Li 77m | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14626, 2712, 289481 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14410, Q9WU65 | |
GK2 | |
Synthetic peptide corresponding to amino acids 276-325 of Human Glycerol kinase 2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction