Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109767
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109767 Supplier Invitrogen™ Supplier No. PA5109767
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAPKTAAVG PLVGAVVQGT NSTRFLVFNS KTAELLSHHK VELTQEFPKE GWVEQDPKEI LQSVYECIAR TCEKLDELNI DISNIKAVGV SNQRETTVIW DKLTGEPLYN AVVWLDLRTQ TTVEDLSKKI PGNSNFVKSK TGLPLSTYFS.

GK2 (glycerol kinase 2), also known as GKP2 or GKTA, is a 553 amino acid protein that belongs to the FGGY kinase family and is involved in the pathway of glycerol degradation. Localized to the outer membrane of the mitochondrion and expressed at high levels in testis, GK2 functions to catalyze the ATPdependent conversion of glycerol to glycerol 3-phosphate. Via its catalytic activity, GK2 plays an essential role in the regulation of glycerol uptake and metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GK2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GK2
Gene Accession No. Q14410, Q9WU65
Gene Alias ATP:glycerol 3-phosphotransferase 2; GK; GK 2; GK2; GKP2; Gkrs2; Gk-rs2; GKTA; glucokinase activity, related sequence 2; Glycerokinase; glycerokinase 2; glycerol kinase 2; glycerol kinase pseudogene 2; glycerol kinase testis specific 2; glycerol kinase, testis specific 2; testis tissue sperm-binding protein Li 77m
Gene Symbols GK2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GK2 (NP_149991.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14626, 2712, 289481
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.