Learn More
Invitrogen™ Glycerol kinase 3 Monoclonal Antibody (2H4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518968
Description
Peptide Sequence: MAASKKAVLG PLVGAVDQGT SSTRFLVFNS RTAELLSHHQ VEIKQEFPRE GWVEQDPKEI LHSVYECIEK TCEKLGQLNI GISNIKAIGV SNQRETTVVW DKITGEPLYN AVVWLDLRTQ STVESLSKRI PGNNNFVKSK TGLPLSTYFS AVKLRWLLDN VRKVQKAVEE KRALFGTIDS WLIWSLTGGV NGGVHCTDVT NASRTMLFNI HSLEWDKQLC EFFGIPMEIL PHVRSSSEIY GLMKAGALEG VPISGCLGDQ SAALVGQMCF QIGQAKNTYG TGCFLLCNTG HKCVFSDHGL LTTVAYKLGR DKPVYYALEG SVAIAGAVIR WLRDNLGIIK TSEEIEKLAK EVGTSYGCYF VPAFSGLYAP YWEPSARGII CGLTQFTNKC HIAFAALEAV CFQTREILDA MNRDCGIPLS HLQVDGGMTS NKILMQLQAD ILYIPVVKPL MPETTALGAA MAAGAAEGVD VWSLEPEDLS AVTMKRFEPQ INAEESEIRY STWKKAVMKS MGWVTTQSPE GGDPSVFCSL PLGFFIVSSM AMLIGARYIS GIP.
Key enzyme in the regulation of glycerol uptake and metabolism.
Specifications
Glycerol kinase 3 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q14409 | |
GK3P | |
GK3P (AAH66960, 1 a.a. ~ 553 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
2H4 | |
Unconjugated | |
GK3P | |
GK3P; GKP3; GKTB; glycerol kinase 3 pseudogene | |
Mouse | |
Affinity chromatography | |
RUO | |
2713 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.