Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glycerol kinase 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549825 Supplier Invitrogen™ Supplier No. PA549825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total Glycerol Kinase 3 protein.

Key enzyme in the regulation of glycerol uptake and metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glycerol kinase 3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GK3P
Gene Accession No. Q14409
Gene Alias GK3P; GKP3; GKTB; glycerol kinase 3 pseudogene
Gene Symbols GK3P
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human GLPK3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2713
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.