Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycerol Kinase Antibody, DyLight 680, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP178018FR
Description
Glycerol Kinase Polyclonal antibody specifically detects Glycerol Kinase in Bacteria samples. It is validated for Western Blot, ELISASpecifications
Glycerol Kinase | |
Polyclonal | |
Western Blot, ELISA | |
ATP:glycerol 3-phosphotransferase, EC 2.7.1.30, GK1, GKD, Glycerokinase, glycerol kinase | |
Glycerol Kinase [Cellulomonas species] (Uniprot: D0VZG4) | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
2710 | |
Store at 4C in the dark. |
Western Blot, ELISA | |
DyLight 680 | |
50mM Sodium Borate | |
Goat | |
Multi-step | |
RUO | |
Primary | |
Bacteria | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction