Learn More
Glycerol Kinase Polyclonal Antibody, HRP, Rockland™

Goat Polyclonal Antibody
$174.73 - $613.35
Specifications
Antigen | Glycerol Kinase |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | HRP |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0358
![]() ![]() |
Rockland Immunochemicals
2001032110100 |
100 μg |
Each for $613.35
|
|
|||||
50-199-0359
![]() ![]() |
Rockland Immunochemicals
200103211S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Glycerol Kinase is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Goat Serum as well as purified and partially purified Glycerol Kinase [Cellulomonas species]. Cross reactivity against Glycerol Kinase from other sources is unknown.
The product of this gene belongs to the FGGY kinase family of proteins and encodes glycerol kinase. Glycerol kinase is a key enzyme in the regulation of glycerol uptake and metabolism. It catalyzes the phosphorylation of glycerol by ATP, yielding ADP and glycerol-3-phosphate. Defects in this gene are the cause of glycerol kinase deficiency (GKD). Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
Glycerol Kinase | |
ELISA, Immunoprecipitation, Western Blot | |
HRP | |
Goat | |
Bacteria | |
ASTP; ATP:glycerol 3-phosphotransferase; ATP-stimulated glucocorticoid-receptor translocation promoter; ATP-stimulated glucocorticoid-receptor translocaton promoter; BOS_26177; D930012N15Rik; EC 2.7.1.30; Gk; GK 3; GK1; GK3P; GKD; GKl; GKP3; GKTB; glycerokinase; Glycerokinase 3; Glycerol kinase; glycerol kinase-like; Glycerolkinase; Gyk; LOC100354726 | |
Primary | |
Antibody |
1 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2 | |
Glycerol Kinase [Cellulomonas species] | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.