Learn More
Invitrogen™ Glycerol kinase Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120997
Description
Positive test controls include: DU145, U-87MG. The target is usually found in the following locations: Cytoplasm, Cytoplasmic side, Mitochondrion outer membrane, Peripheral membrane protein. Immunogen sequence: GIPLSHLQVD GGMTSNKILM QLQADILYIP VVKPSMPETT ALGAAMAAGA AEGVGVWSLE PEDLSAVTME RFEPQINAEE SEIRYSTWKK AVMKSMGWVT TQSPESGDPS IFCSLPLGFF IVSSMVMLIG ARYISGIP.
The product of this gene belongs to the FGGY kinase family of proteins and encodes glycerol kinase. Glycerol kinase is a key enzyme in the regulation of glycerol uptake and metabolism. It catalyzes the phosphorylation of glycerol by ATP, yielding ADP and glycerol-3-phosphate. Defects in this gene are the cause of glycerol kinase deficiency (GKD). Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| Glycerol kinase | |
| Polyclonal | |
| Unconjugated | |
| GK | |
| ASTP; ATP:glycerol 3-phosphotransferase; ATP-stimulated glucocorticoid-receptor translocation promoter; ATP-stimulated glucocorticoid-receptor translocaton promoter; D930012N15Rik; EC 2.7.1.30; GK; GK 3; GK1; GK3P; GKD; GKl; GKP3; GKTB; glycerokinase; Glycerokinase 3; glycerol kinase; Glycerolkinase; Gyk | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14933, 2710, 79223 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.04 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P32189, Q63060, Q64516 | |
| GK | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 422-559 of Glycerol kinase (GK) (NP_001191948.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.