Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycerol kinase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550970
Description
Glycerol kinase Polyclonal Antibody detects endogenous levels of Glycerol kinase protein.
Glycerol kinase Polyclonal antibody specifically detects Glycerol kinase in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Glycerol kinase | |
Polyclonal | |
Unconjugated | |
GK | |
ASTP; ATP:glycerol 3-phosphotransferase; ATP-stimulated glucocorticoid-receptor translocation promoter; ATP-stimulated glucocorticoid-receptor translocaton promoter; D930012N15Rik; EC 2.7.1.30; GK; GK 3; GK1; GK3P; GKD; GKl; GKP3; GKTB; glycerokinase; Glycerokinase 3; glycerol kinase; Glycerolkinase; Gyk | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14933, 2710, 79223 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P32189, Q63060, Q64516 | |
GK | |
Synthesized peptide derived from the C-terminal region of human GK1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction