Learn More
Glycerol KinaseGoat anti-Bacteria, Polyclonal, Rockland™

Goat Polyclonal Antibody
Supplier: Rockland Immunochemicals 100101211

Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-goat serum, purified and partially purified Glycerol Kinase [Cellulomonas species]. Cross reactivity against Glycerol Kinase from other sources is unknown.
The product of this gene belongs to the FGGY kinase family of proteins and encodes glycerol kinase. Glycerol kinase is a key enzyme in the regulation of glycerol uptake and metabolism. It catalyzes the phosphorylation of glycerol by ATP, yielding ADP and glycerol-3-phosphate. Defects in this gene are the cause of glycerol kinase deficiency (GKD). Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
Glycerol Kinase | |
Polyclonal | |
Unconjugated | |
ASTP; ATP:glycerol 3-phosphotransferase; ATP-stimulated glucocorticoid-receptor translocation promoter; ATP-stimulated glucocorticoid-receptor translocaton promoter; BOS_26177; D930012N15Rik; EC 2.7.1.30; Gk; GK 3; GK1; GK3P; GKD; GKl; GKP3; GKTB; glycerokinase; Glycerokinase 3; Glycerol kinase; glycerol kinase-like; Glycerolkinase; Gyk; LOC100354726 | |
Glycerol Kinase [Cellulomonas species] | |
RUO | |
Bacteria | |
Antibody |
ELISA, Immunoprecipitation, Western Blot | |
90 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Goat | |
2 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.