Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycerol monoacetate, tech., mixture of isomers, cont. varying amounts of diacetate
Click to view available options
Quantity:
250 g
1000 g
Description
Glycerol monoacetate is used for production of tanning leather and dye. It is also used as a solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycerol monoacetate is used for production of tanning leather and dye. It is also used as a solvent.
Solubility
Soluble in water, methanol, ethanol, ether (slightly), and insoluble in benzene.
Notes
Hygroscopic. Protect from moisture. Store at room temperature. Store away from strong oxidizing agents.
Glycerol monoacetate is used for production of tanning leather and dye. It is also used as a solvent.
Solubility
Soluble in water, methanol, ethanol, ether (slightly), and insoluble in benzene.
Notes
Hygroscopic. Protect from moisture. Store at room temperature. Store away from strong oxidizing agents.

Specifications
Specifications
Density | 1.21 |
Boiling Point | 258°C |
Flash Point | 145°C (293°F) |
Refractive Index | 1.45 |
Quantity | 1000 g |
Sensitivity | Hygroscopic |
Merck Index | 14,6242 |
Solubility Information | Soluble in water,methanol,ethanol,ether (slightly),and insoluble in benzene. |
Formula Weight | 134.13 |
Grade | Technical |
Show More |
Safety and Handling
EINECSNumber | 247-704-6 |
RTECSNumber | AK3595000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction