Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycerol ethoxylate-co-propoxylate triol

Catalog No. 409189500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
500 mL

Catalog No. 40-918-9500ML

Supplier: Sigma Aldrich 409189500ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Glycerol ethoxylate-co-propoxylate triol is a cross-linker that can be used as a UV curable binder for polyurethane acrylate. It has a low viscosity that makes it useful in the preparation of inks for ink-jet printing and the UV based drying of these pigments.

Specifications

CAS 51258-15-2
Density 1.063 g/mL (at 25°C)
Refractive Index n20/D 1.462 (literature)
Linear Formula HO(C3H6O)x(CH2CH2O)yCH[CH2(OCH2CH2)y(OC3H6)xOH]2
MDL Number MFCD00197863
Quantity 500 mL
Molecular Weight (g/mol) 2600
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.