Learn More
Glycerol, puriss., Meets analytical specification of Ph. Eur. BP, USP, FCC., Solstice
Shop All Honeywell Research Chemicals ProductsDescription
- Solstice has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Melting Point | 18°C |
| Color | Colorless |
| Density | 1.2580 to 1.2644g/mL at 20°C, ≥1.257g/mL at 25°C |
| Boiling Point | 290°C |
| Flash Point | 160°C (320°F) |
| Ignition Residue | ≤0.01% (as SO4) |
| Ignition Point | 698°F |
| Packaging | Poly Drum |
| Refractive Index | n20/D 1.474(lit.) |
| Linear Formula | HOCH2CH(OH)CH2OH |
| Show More |
Safety and Handling
| EINECSNumber | 200-289-5 |
| RTECSNumber | MA8050000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.