Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, 10% (w/v), Ricca Chemical
SDP

Catalog No. 328332 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
30 gal.
Packaging:
Natural Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3283-32 1 L Natural Poly Bottle
32834 120 mL Natural Poly Bottle
3283-16 500 mL Natural Poly Bottle
328330 30 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 3283-32 Supplier Ricca Chemical Company Supplier No. 328332
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56-40-6 , 7732-18-5
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Color Colorless
CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 4
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Glycine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.