Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, MP Biomedicals™

Catalog No. p-4495716
Click to view available options
Quantity:
50 g
250 g
1 kg
3 kg

Glycine, ACS Reagent Grade, White Crystalline Powder

  • It is widely used as a buffer for a variety of immunological applications.
  • It is also used as an eluent in column chromatography.
  • It has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.
  • Commonly used as a component in Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis, a component of Towbin's transfer buffer for Western blots , a buffer substance in cryoenzymology, in osmotic pressure maintenance in isoelectric focusing of erythrocytes, salting-in effect in protein chemistry, and as a buffer component in the coupled phosphatase-kinase reaction for end labelling of restriction fragments.
  • Glycine is a non-essential amino acid. Only amino acid with no asymmetric carbon.
  • It is a major inhibitory neurotransmitter.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 240°C (Decomposition)(Literature)
Color White
Assay Percent Range ≥98.5%
Beilstein 635782
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
Solubility Information Soluble in water (250mg/mL at 25°C; 672mg/mL at 100°C); slightly soluble in alcohol and ethanol(95%); almost insoluble in ether.
Density 1.16g/mL (Lit.)
Quantity 50 g
Formula Weight 75.07
Percent Purity ≥98.5%
Grade Reagent
Physical Form Crystalline Powder
Assay ≥98.5%
Chemical Name or Material Glycine, For ACS analysis
Show More Show Less
RTECSNumber MB7600000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.