Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, 98%, extra pure, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
25kg
56-40-6
C2H5NO2
75.067
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
56-40-6
C2H5NO2
75.067
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
Arsenic (As) 3ppm max.
Heavy Metals (as Pb) 20ppm max.
Assay Percent Range 98%
Linear Formula H2NCH2CO2H
Infrared Spectrum Authentic
Beilstein 04, 333
Fieser 01,412
Merck Index 15, 4526
Loss on Drying 0.2% max. (105°C, 2 hrs)
Packaging Plastic bucket
Residue after Ignition 0.1% max.
Formula Weight 75.07
Physical Form Liquid
Percent Purity ≥97.5% (Total base)
Grade Extra Pure
Density 1.5950g/mL
Color Colorless to Yellow
pH 5.5 to 6.5 (5% aq. soln.)
Quantity 25kg
Chemical Name or Material Glycine
Show More Show Less