Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Glycine, 99+%, for analysis

Catalog No. AC220910010
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Plastic Bottle
Plastic Drum

Glycine, CAS # 56-40-6, is a purified formulation of the amino acid, glycine, (aminoacetic acid) that is used in the synthesis of several compounds including creatinine, purines and porphyrins. | CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Glycine is an essential acid that can be used for synthesis of creatine, purines and porphyrins
  • Soluble in water; has a pH of ∼=5.5-6.5 in solution
  • Glycine is an inhibitory neurotransmitter that regulates the activity of NMDA receptors and affects neural activity

Applications

  • Glycine may also be used to maintain osmotic pressure, or as a component of other molecular biology buffers that are ion-dependent
  • Glycine is used in molecular biology as a buffer component for gel electrophoresis; the ability of glycine to adopt a negative charge at higher pH makes it ideal for separating proteins in the stacking layer
  • Intended for biochemistry applications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Ammonium 200ppm max.
Arsenic (As) 1ppm max.
Color White
Chlorine (Cl) 40ppm max.
Hydrolyzable Substance Clear solution
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.2% (105°C, 2 hrs) max.
Assay Percent Range 99+%
Packaging Plastic Bottle
Linear Formula H2NCH2CO2H
Residual Solvents Meets the requirements
Sulfate 50ppm max.
Sulfated Ash 0.1% max.
Density 1.5950g/mL
Quantity 1 kg
Formula Weight 75.07
Percent Purity ≥99.0% (on dried substance) (total base)
Grade Analytical
Physical Form Crystalline Powder
Chemical Name or Material Glycine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.