Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, ≥99%, Molecular Biology Grade, Ultrapure

Catalog No. aaj1640736
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle

CAS: 56-40-6 | C2H5NO2 | 75.067 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.067
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES C(C(=O)O)N

Specifications

Melting Point ∼245°C (decomposition)
Packaging Plastic Bottle
Linear Formula H2NCH2CO2H
Beilstein 635782
Density 1.595
Quantity 500 g
Formula Weight 75.07
Percent Purity ≥99%
Grade Molecular Biology
Specific Gravity 1.595
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less
EINECSNumber 200-272-2
RTECSNumber MB7600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.