Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, BAKER ANALYZED™ Biochemical Reagent, J.T. Baker™

Catalog No. p-5391632
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle

High quality biochemicals for research use

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 1 kg
Formula Weight 75.07
Grade BAKER ANALYZED™ Biochemical Reagent
Specific Gravity 1L = 1.16kg
Chemical Name or Material Glycine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.