Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

Click to view available options
Quantity:
1kg
5kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle

Chemical Identifiers

56-40-6
C2H5NO2
75.067
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
CHEBI:15428

High quality biochemicals for research use

CAS 56-40-6
Molecular Formula C2H5NO2
Packaging Wide Mouth Amber Glass Bottle
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILES C(C(=O)O)N
IUPAC Name 2-aminoacetic acid
Molecular Weight (g/mol) 75.067
PubChem CID 750
ChEBI CHEBI:15428
Formula Weight 75.07
Quantity 1kg
Specific Gravity 1L = 1.16kg
Chemical Name or Material Glycine
Grade BAKER ANALYZED™ Biochemical Reagent
Show More Show Less