Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. B026625G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1738-76-7
Molecular Formula C16H19NO5S
Molecular Weight (g/mol) 337.39
MDL Number MFCD00035425
InChI Key WJKJXKRHMUXQSL-UHFFFAOYSA-N
Synonym h-gly-obzl.tosoh, glycine benzyl ester p-toluenesulfonate, glycine benzyl ester p-toluenesulfonate salt, benzyl glycinate p-toluenesulfonate, o-benzylglycine toluene-p-sulphonate, h-gly-obzl p-tosylate, gly-obzl.tsoh, aminoacetic acid benzyl ester toluene-4-sulfonic acid salt, glycine benzyl ester p-tosylate, glycine benzyl ester 4-toluenesulfonate
PubChem CID 6451311
IUPAC Name benzyl 2-aminoacetate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CN

Specifications

Melting Point 135°C
Color White
Quantity 25 g
Formula Weight 337.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Glycine Benzyl Ester p-Toluenesulfonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.