Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, MP Biomedicals™

Catalog No. MP21005705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21005705 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21005705 Supplier MP Biomedicals, Inc Supplier No. 02100570.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be synthesized in the body by Serine Hydroxymethyltransferase. Glycine, MP Biomedicals is a nonessential, non-chiral amino acid that acts as an inhibitory neurotransmitter, particularly in brain stem and medulla. It is commonly used as a buffer compon

Glycine does not have a chiral center, so it is the only amino acid with no asymmetric carbon. Therefore, it does not have a D- or L- isomer designation. This material is synthetic. It is the smallest of the 20 natural amino acids. It acs as inhibitory neurotransmitter that is shown to block NMDA-stimulated dopamine release in fetal mesencephalic cell cultures. It is also reported to increase NMDA-receptor mediated synaptic potential in cortical neurons.

Glycine is commonly used as a component in:

  • Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis
  • Towbin's transfer buffer for Western blots
  • Buffers used in cryoenzymology, osmotic pressure maintenance in isoelectric focusing of erythrocytes and salting-in effect in protein chemistry
  • Buffer for coupled phosphatase-kinase reaction for end labelling of restriction fragments
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Solubility Information Soluble in water (250mg/mL at 25°C; 672mg/mL at 100°C); slightly soluble in alcohol and ethanol (95%); almost insoluble in ether.
Quantity 500 g
Formula Weight 75.07
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Glycine
Recommended Storage Store at Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.