Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycine, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500g
2kg
Packaging:
Wide Mouth Amber Glass Bottle
56-40-6
C2H5NO2
75.067
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
56-40-6
C2H5NO2
75.067
DHMQDGOQFOQNFH-UHFFFAOYSA-N
glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine
750
CHEBI:15428
2-aminoacetic acid
C(C(=O)O)N
Packaging Wide Mouth Amber Glass Bottle
Formula Weight 75.07
Grade Multi-Compendial/USP
Quantity 2kg
Specific Gravity 1L = 1.16kg
Chemical Name or Material Glycine