Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycine N-Methyltransferase/GNMT Antibody (5D1), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259452AF350
This item is not returnable.
View return policy
Description
Glycine N-Methyltransferase/GNMT Monoclonal antibody specifically detects Glycine N-Methyltransferase/GNMT in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Glycine N-Methyltransferase/GNMT | |
Monoclonal | |
Alexa Fluor 350 | |
EC 2.1.1.20, glycine N-methyltransferase | |
Recombinant human Glycine N-Methyltransferase/GNMT (1-295aa) purified from E. coli | |
0.1 mL | |
Vision | |
27232 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA, Immunofluorescence | |
5D1 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction