Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycine N-Methyltransferase/GNMT Antibody (OTI8A3), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271838AF405
Description
Glycine N-Methyltransferase/GNMT Monoclonal specifically detects Glycine N-Methyltransferase/GNMT in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
Glycine N-Methyltransferase/GNMT | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GNMT | |
Full length recombinant protein of human Glycine N-Methyltransferase/GNMT (NP_061833) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG3 |
Western Blot, Flow Cytometry | |
OTI8A3 | |
Western Blot, Flow Cytometry | |
EC 2.1.1.20, glycine N-methyltransferase | |
Mouse | |
Affinity Purified | |
Vision | |
27232.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction