Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycine N-Methyltransferase/GNMT Antibody (OTI8A3), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271838APC
Description
Glycine N-Methyltransferase/GNMT Monoclonal specifically detects Glycine N-Methyltransferase/GNMT in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| Glycine N-Methyltransferase/GNMT | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| GNMT | |
| Full length recombinant protein of human Glycine N-Methyltransferase/GNMT (NP_061833) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG3 |
| Flow Cytometry | |
| OTI8A3 | |
| Flow Cytometry | |
| EC 2.1.1.20, glycine N-methyltransferase | |
| Mouse | |
| Affinity Purified | |
| Vision | |
| 27232.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction