Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Glycine N-Methyltransferase/GNMT Antibody (OTI8A3), DyLight 550, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271838R
Description
Glycine N-Methyltransferase/GNMT Monoclonal specifically detects Glycine N-Methyltransferase/GNMT in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
| Glycine N-Methyltransferase/GNMT | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GNMT | |
| Full length recombinant protein of human Glycine N-Methyltransferase/GNMT (NP_061833) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG3 | 
| Western Blot, Flow Cytometry | |
| OTI8A3 | |
| Western Blot, Flow Cytometry | |
| EC 2.1.1.20, glycine N-methyltransferase | |
| Mouse | |
| Affinity Purified | |
| Vision | |
| 27232.0 | |
| Store at 4°C in the dark. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction