Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycine N-Methyltransferase/GNMT Antibody (OTI8A3), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271838MFV500
Description
Glycine N-Methyltransferase/GNMT Monoclonal antibody specifically detects Glycine N-Methyltransferase/GNMT in Human, Mouse samples. It is validated for Flow CytometrySpecifications
Glycine N-Methyltransferase/GNMT | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
OTI8A3 | |
Flow Cytometry | |
EC 2.1.1.20, glycine N-methyltransferase | |
Full length human recombinant protein of human GNMT (NP_061833) produced in HEK293T cell. | |
0.1 mL | |
Vision | |
27232 | |
Store at 4C in the dark. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction