Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycine, Ultrapure Bioreagent, J.T. Baker™
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
Description
- For Electrophoresis, Liquid Chromatography and Molecular Biology Applications
Specifications
Specifications
Packaging | Poly Bottle |
Quantity | 1 kg |
Formula Weight | 75.07 |
Grade | Ultrapure Bioreagent |
Specific Gravity | 1L = 1.16kg |
Chemical Name or Material | Glycine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction