Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycine, USP, 98.5-101.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Glycine, USP is a colorless, sweet-tasting crystalline non-essential amino acid. USP glycine has a wide variety of uses, including as an additive in pet food and animal feed, in foods and pharmaceuticals as a sweetener/taste enhancer, or as a component of food supplements and protein drinks. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 100% |
Chloride | 0.007% |
Ignition Residue | 0.1% |
Loss on Drying | 0.2% |
Packaging | Poly Bottle |
Sulfate | 0.0065% |
Quantity | 2.5 kg |
Percent Purity | 98.5 to 101.5% |
Grade | USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction